Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ Para-Pak™ Macro-CON™ Stool Concentration System

Catalog No. 14910133 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
120/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-910133 120/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-910133 Supplier Meridian Bioscience™ Supplier No. 970120
This item is not returnable. View return policy

Includes

120 conical centrifuge tubes (50mL), filtration units and screw caps, one bottle surfactant (30mL) and one package insert

Improves small population sampling variation

  • Closed system for the concentration and recovery of eggs, larvae and protozoa from preserved fecal specimens
  • Consists of a separate concentrator/filtration unit to be attached to a Para-Pak Formalin, SAF or EcoFix™ patient specimen collection vial
  • Minimizes potential user contamination and specimen exposure
  • Macro-Con uses the entire contents of the specimen collection vial, reducing variation due to specimen sampling
  • Design enhances organism recovery from stool sample

Specifications

Product Type Stool Concentration System
Certifications/Compliance CE
Includes 50mL Conical centrifuge tubes, filtration units and screw caps, 30mL bottle surfactant and one package insert
Quantity 120/Cs.
Storage Requirements 20°C to 30°C
For Use With (Application) Preserved and unpreserved stool
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.