Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ Para-Pak™ Parasitology Transport Systems Reagents: Ethyl Acetate
GSA_VA

Catalog No. 23312021 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-312021 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-312021 Supplier Meridian Bioscience™ Supplier No. FK4318
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fixatives, stain, and other reagents for use with Meridian Bioscience Para-Pak Parasitology Transport Systems

225mL bottle

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Stool
Certifications/Compliance CE
Color Clear
Format Liquid
Media Type 99.5% Ethylacetate
Packaging Bottle
Quantity 1 Ea.
Storage Requirements 20°C to 30°C
Volume (English) 7.6 oz.
Volume (Metric) 225 mL
For Use With (Application) Stool
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.