Learn More
Meridian Bioscience™ Para-Pak™ Parasitology Transport Systems Reagents: Hemo-De
Shop All Meridian Bioscience ProductsSpecifications
Specifications
| Product Type | Hemo-De Reagent |
| Certifications/Compliance | CE |
| Color | Clear |
| Format | Liquid |
| Media Type | 99% D-Limonene, 1% Butylated Hydroxyanisole |
| Packaging | Bottle |
| Quantity | 1 Ea. |
| Storage Requirements | 15°C to 30°C |
| Volume (English) | 13.5 oz. |
| Volume (Metric) | 400 mL |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.