Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ Para-Pak™ Parasitology Transport Systems Reagents: Wheatley's Trichrome Stain
GSA_VA

Catalog No. 23244633 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
12/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-244633 12/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-244633 Supplier Meridian Bioscience™ Supplier No. 400112
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fixatives, stain, and other reagents for use with Meridian Bioscience Para-Pak Parasitology Transport Systems

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Trichrome Stain
Certifications/Compliance CE
Color Dark Purple
Format Liquid
Media Type 1.02% Acetic Acid, 0.59% Disodium 4,5-dihydroxy-3-phenylazonaphthalene-2,7-disulphonate, 0.68% Tungsten Hydroxide Oxide Phosphate, 0.15% Fast Green fcf, 0.15% Light Green SF
Packaging Bottle
Quantity 12/Cs.
Storage Requirements 15°C to 30°C
Volume (English) 16.9 oz.
Volume (Metric) 500 mL
For Use With (Application) Stool
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.