Learn More
Meridian Bioscience™ Para-Pak™ Parasitology Transport Systems: Three-Vial Kits
Shop All Meridian Bioscience ProductsDescription
Includes
Patient instructions. Three-vial kits sytems are packaged in biobags with illustrated, multi-lingual patient instructions to ensure safe and sanitary specimen collection.
- Choice of fixatives and preservatives
- Each unbreakable vial has a color-coded cap and label, a clearly marked “fill to here” line and a built-in collection spoon in the screw-cap top
10% Formalin Fixative (Neutral Buffered)
- Preserves fecal specimens for concentration and direct examination
- Used for the recovery of enteric bacteria, such as Salmonella, Shigella, Campylobacter and Yersinia
- Medium includes a pH indicator that detects acidic conditions for optimal enteric pathogen recovery
- A 30mL leakproof vial for specimens that do not need a transport medium
- Maintains the bacterial population of enteric pathogens such as Yersinia, Salmonella, Shigella, Campylobacter and Vibrio Cholerae for up to 96 hours
- For the routine collection, transportation, preservation and examination of stool specimens for intestinal parasites
- Combines the fixative properties of formaldehyde, alcohol and merthiolate to maintain the morphology for all stages in the life cycles of intestinal parasites
- Acts as a preservative/fixative for protozoan trophozoites
- Slides fixed with Modified PVA may be stained using trichrome or hematoxyli
- Contains copper sulfate to eliminate disposal problems
- Multi-purpose fixative/preservative suitable for direct examination, concentration and permanent stain from the same specimen
- A mercury-free fixative for permanent staining
- A low-viscosity formulation to preserve fecal specimens for permanent staining procedures
- Fast penetration and drying formula provides excellent fixation
Specifications
Specifications
| Product Type | Parasitology Transport System |
| Certifications/Compliance | CE |
| Includes | Patient instructions, three vial kits sytems are packaged in biobags with illustrated, multi-lingual patient instructions |
| Media Type | LV-PVA, 10% Formalin, Clean |
| No. of Vials | 3.0 |
| Quantity | 7 Pk. |
| Storage Requirements | 20°C to 30°C |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.