Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ Para-Pak™ Parasitology Transport Systems: Two-Vial Kits
GSA_VA

Catalog No. 14910143 Shop All Meridian Bioscience Products
Change view
Click to view available options
Media Type:
LV-PVA, 10% Formalin
Modified (Cu) PVA, 10% Formalin
SAF, Clean
Zn-PVA, 10% Formalin
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Media Type
14-910-143 SAF, Clean
14-910-201 LV-PVA, 10% Formalin
14-910-135 Modified (Cu) PVA, 10% Formalin
14-910-151 Zn-PVA, 10% Formalin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 14-910-143 Supplier Meridian Bioscience™ Supplier No. 300212
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Patient instructions. Two-vial kits sytems are packaged in biobags with illustrated, multi-lingual patient instructions to ensure safe and sanitary specimen collection.

Vials and vial kits designed to improve the overall processing of specimens for examination of ova, parasites and other enteric pathogens

  • Choice of fixatives and preservatives
  • Each unbreakable vial has a color-coded cap and label, a clearly marked “fill to here” line and a built-in collection spoon in the screw-cap top

10% Formalin Fixative (Neutral Buffered)

  • Preserves fecal specimens for concentration and direct examination
  • Culture and Sensitivity (C & S) Media
    • Used for the recovery of enteric bacteria, such as Salmonella, Shigella, Campylobacter and Yersinia
    • Medium includes a pH indicator that detects acidic conditions for optimal enteric pathogen recovery
    Clean Vials
    • A 30mL leakproof vial for specimens that do not need a transport medium
  • Enteric Plus (Cary-Blair) Media
    • Maintains the bacterial population of enteric pathogens such as Yersinia, Salmonella, Shigella, Campylobacter and Vibrio Cholerae for up to 96 hours
  • MF (Formaldehyde, Alcohol and Merthiolate) Fixative
    • For the routine collection, transportation, preservation and examination of stool specimens for intestinal parasites
    • Combines the fixative properties of formaldehyde, alcohol and merthiolate to maintain the morphology for all stages in the life cycles of intestinal parasites
    Modified Cupric Sulfate–Polyvinyl Alcohol (Modified Cu–PVA)
    • Acts as a preservative/fixative for protozoan trophozoites
    • Slides fixed with Modified PVA may be stained using trichrome or hematoxyli
    • Contains copper sulfate to eliminate disposal problems
    Sodium Acetate Acetic Acid Formalin (SAF)
    • Multi-purpose fixative/preservative suitable for direct examination, concentration and permanent stain from the same specimen
  • Zinc Sulfite–Polyvinyl Alcohol (Zn–PVA)
    • A mercury-free fixative for permanent staining
  • Low Viscosity–Polyvinyl Alcohol (LV–PVA)
    • A low-viscosity formulation to preserve fecal specimens for permanent staining procedures
    • Fast penetration and drying formula provides excellent fixation

    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

    Specifications

    Product Type Parasitology Transport System
    Certifications/Compliance CE
    Format Vial
    Includes Patient instructions, two vial kits sytems are packaged in biobags with illustrated, multi-lingual patient instructions
    Media Type SAF, Clean
    No. of Vials 2.0
    Quantity 120 Sets/Case
    Storage Requirements 20°C to 30°C
    WARNING: Reproductive Harm - www.P65Warnings.ca.gov
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.