Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ Premier™ C. difficile GDH
GSA_VA

Catalog No. 23029255 Shop All Meridian Bioscience Products
Change view
Click to view available options
Detectable Analytes:
C. difficile GDH Antigen
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
23-029-255 C. difficile GDH Antigen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-029-255 Supplier Meridian Bioscience™ Supplier No. 611096
Add to Cart
Add to Cart
This item is not returnable. View return policy

Enzyme immunoassay for detection of Clostridium difficile common antigen (GDH)

  • Early and accurate elimination of negative results with results within one hour.
  • Detection in fecal specimens from symptomatic persons suspected of having C. difficile infection (CDI).
  • No differentiation between toxigenic and non-toxigenic strains of C. difficile.
  • Shelf Life is 18 months from the date of manufacture.
  • Sensitivity: 94%.
  • Specificity: 95%.

Certifications

CE marked

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance CE Marked
Detectable Analytes C. difficile GDH Antigen
Type Clostridium difficile GDH Enzyme Immunoassay
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.