Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ Premier™ H. pylori Microplate Assay
GSA_VA

Catalog No. 14910164 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-910-164 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-910-164 Supplier Meridian Bioscience™ Supplier No. 606096
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Results require no calculations

  • CLIA Complexity: Non-waived
  • CPT Code: 87338
  • Microwell EIA for detection of IgG antibodies specific to Helicobacter pylori in serum and plasma
  • Results in 50 minutes; results read visually or spectrophotmetrically
  • Microwell-based test uses a sonicated bacterial cell lysate as the capture antigen
  • Sensitivity: 99%; Specificity: 96%

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Content And Storage 2°C to 8°C
Clia Complexity Non-waived
Detectable Analytes H. pylori IgG
Format Microwell
Sample Type Plasma, Serum
Sensitivity 99.2%
Type H. pylori IgG Test Kit
Quantity 96 Tests
Specificity 0.96
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.