Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ TRU™ Legionella Test Kit
GSA_VA

Catalog No. 23043201 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
32 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-043-201 32 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-043-201 Supplier Meridian Bioscience™ Supplier No. 751930
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rapid assay for detection of Legionella pneumophila Serogroup-1

  • Lateral flow assay of urine specimens complies with CDC recommendations for Legionella pneumophila sg1
  • Positive agreement 95.5%; Negative agreement 100%
  • Outcome in 20 minutes
  • Three-step procedure
  • Simple interpretive results: Negative test result shows only control line; Positive test shows both control line and test line

Certifications

CE marked

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

CPT Code 87449
Sample Type Boric Acid Preserved Urine, Fresh Urine
Type TRU™ Legionella Test Kit
For Use With (Application) Detection of Legionella pneumophila serogroup-1 and lateral flow test of Urine specimens
Quantity 32 Tests
Test Time 20 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.