Learn More
Meridian Bioscience™ TRU RSV™ Tests
GSA_VA
Catalog No. 23030701 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
32 Tests
Catalog No. Quantity
23-030-701 32 Tests
1 options
This item is not returnable. View return policy
Catalog No. 23-030-701 Supplier Meridian Bioscience™ Supplier No. 751330
Add to Cart
Add to Cart
This item is not returnable. View return policy
Test under the hood without concern about airflow
  • One-step immunochromatographic assay
  • For rapid antigen detection
  • Use Pooled Monoclonal Antibodies detecting two RSV proteins which may help detect more positive samples
  • Easy-to-use controls are validated with both TRU RSV and TRU FLU™
Limit pathogen exposure in the laboratory
  • Test design ideal for testing under the hood
  • Closed immediately after patient sample addition, and remains closed for interpretation and disposal
Optimize your lab workflow
  • Convenient workstation supplied allows six tests to be performed in the space of one traditional card test
  • Tests are easily batched
  • Save time with easy, ready-to-use external controls
Limit susceptibility to variation in operator technique
  • Easy, single-step sample addition
  • Detects both the fusion protein and the nucleoprotein antigens of the RSV virus
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
CPT Code 87807
Detectable Analytes Respiratory Syncytial Virus
Sample Type Nasal Aspirate, Nasal Swab, Nasal Wash
Type RSV Test Kit
Includes Test strip, conjugate tube, sample diluent/negative control, Plastic pipettes
Quantity 32 Tests
Test Time 15 min.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov