Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ Latex Agglutination System
GSA_VA

Catalog No. 23024866 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
50 to 100 Determinations
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-024866 50 to 100 Determinations
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-024866 Supplier Meridian Bioscience™ Supplier No. 140100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Sample diluent, Detection latex, Control latex, Antibody control, Negative control, Positive control, Pronase, Disposable reaction cards, Reaction reference photograph, Package insert

Requires

Purified water, 1 x 0.01mL pipettes, Waterbath or heat block, Small serologic test tubes, Rack, Marking pen, Applicator sticks, 25μL/ 100μL/ 200μL (or equivalent) pipetter

Qualitative and semi-quantitative latex test for detection of Cryptococcus neoformans

  • Detects capsular polysaccharide antigens of Cyptococcus neoformans in serum and cerebrospinal fluid
  • 50 to 100 determinations

Certifications

CE marked

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Crypto coccal Antigen latex agglutination system
Quantity 50 to 100 Determinations
Sample Type Serum, CSF
Content And Storage 2°C to 8°C
Sufficient For 50 to 100 Determinations
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.