Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ ImmunoCard™ Helicobacter pylori Test Kit
GSA_VA

Catalog No. 14910105 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
30 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-910-105 30 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-910-105 Supplier Meridian Bioscience™ Supplier No. 710030
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Test cards, positive and negative control, enzyme conjugate, wash buffer, substrate reagent, whole blood sample diluent, and transfer pipets.

Easily convertible to a batchable system as test volume increases

  • CPT Code: 87338
  • CLIA Complexity: Non-waived
  • Rapid EIA
  • Test results in six minutes
  • Noninvasive
  • Detects IgG for Helicobacter pylori in serum or plasma
  • Simple, three-step procedure

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Content And Storage 2°C to 8°C
Clia Complexity Non-waived
Detectable Analytes H. pylori IgG
Format Test Card
Sample Type Plasma, Serum, Whole Blood
Sensitivity 97%
Type H. pylori Test Kit
Includes Test cards, positive and negative control, Enzyme conjugate, Wash Buffer, substrate reagent, whole Blood sample diluent and transfer pipets
Quantity 30 Tests
Specificity 0.981
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.