Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ ImmunoCard Mycoplasma Test Kit
GSA_VA

Catalog No. 14910108 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
30 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-910-108 30 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-910-108 Supplier Meridian Bioscience™ Supplier No. 709030
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

all materials, including positive and negative controls for 30 determinations

Simple four-step procedure

  • Test uses reliable EIA membrane technology
  • Detects IgM antibodies to Mycoplasma pneumoniae in human serum
  • Results in less than ten minutes

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Content And Storage Refrigerate
Detectable Analytes M. pneumoniae IgM
Sample Type Human Serum
Sensitivity 88%
Type ImmunoCard™ Mycoplasma Test Kit
Includes Positive and negative control
Quantity 30 Tests
Specificity 0.9
Test Time 20 min.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.