Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ ImmunoCard™ STAT! Cryptosporidium/Giardia Test Kit
GSA_VA

Catalog No. 14910190 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
30 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-910-190 30 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-910-190 Supplier Meridian Bioscience™ Supplier No. 750830
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Kits contain buffer, sample tube, conjugates A and B, and test devices; Positive controls contain 1mL Giardia and 1mL Crypto Positive Control vials (16 determinations each).

One-step test for direct detection of Cryptosporidium and Giardia lamblia

  • CLIA Complexity: Non-waived
  • CPT Codes: 87328, 87329
  • Sensitivity: 97%
  • Specificity: 100% for both organisms
  • 10-minute test
  • Monoclonal antibody technology distinguishes between Cryptosporidium and Giardia lamblia
  • Qualitative results for Cryptosporidium parvuum and Giardia lamblia
  • In vitro diagnostic use as an aid in detection of suspected Cryptosporidium or Giardia infections

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Content And Storage 2°C to 8°C
CPT Code 87328, 87329
Description ImmunoCard Stat™ Cryptosporidium/Giardia Test Kit
Clia Complexity Non-waived
Detectable Analytes Cryptosporidium, G. lamblia
Sensitivity 97%
Type ImmunoCard Stat™ Cryptosporidium/Giardia Test Kit
Includes Buffer, sample tube, conjugates A and B and test devices
Quantity 30 Tests
Specificity 1
Test Time 10 min.
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.