Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ ImmunoCard™ STAT! Cryptosporidium/Giardia Test: Controls
GSA_VA

Catalog No. 23030870 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
16 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-030-870 16 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-030-870 Supplier Meridian Bioscience™ Supplier No. 750820
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Controls for use with the Meridian ImmunoCard STAT Cryptosporidium/Giardia

  • CLIA Complexity: Non-waived
  • CPT Codes: 87328, 87329
  • Sensitivity: 97%
  • Specificity: 100% for both organisms
  • 10-minute test
  • Monoclonal antibody technology distinguishes between Cryptosporidium and Giardia lamblia
  • Qualitative results for Cryptosporidium parvuum and Giardia lamblia
  • In vitro diagnostic use as an aid in detection of suspected Cryptosporidium or Giardia infections

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

CPT Code 87328, 87329
Description Positive Control
Clia Complexity Non-waived
Detectable Analytes Cryptosporidium, G. lamblia
Format Vial
Type Positive Control
Includes 1mL Giardia and 1mL Crypto positive control vials
Quantity 16 Tests
Test Time 10 min.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.