Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ ImmunoCard™ STAT! E. coli 0157 Plus Test Kit
GSA_VA

Catalog No. 14910305 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
30 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-910-305 30 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-910-305 Supplier Meridian Bioscience™ Supplier No. 750530
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Each kit contains individually foil-wrapped test devices, positive and negative controls, sample diluent and 30 transfer pipets

Testing can be performed directly on stool samples or on stool culture colonies

  • CLIA Complexity: Non-waived
  • CPT Code: 87046
  • Sensitivity: With direct stool sample, 82%; with samples from stool culture enrichments or stool culture plate, 100%
  • Specificity: With direct stool sample, 99%; with samples from stool culture enrichments or stool culture plate, 100%
  • Direct antigen rapid immunoassay for the detection of Shiga toxin-producing E. coli 0157
  • Direct qualitative results within 10 minutes
  • Results interpreted visually by the presence or absence of a colored line

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Content And Storage 2°C to 8°C
Clia Complexity Non-waived
Detectable Analytes E. coli Serogroup O157
Sensitivity 82% Stool, 100% Isolates
Type ImmunoCard Stat™ E. coli 0157 Plus Test Kit
Includes Foil-wrapped test devices, positive and negative control, sample diluent and 30 transfer pipets
Quantity 30 Tests
Specificity 99% stool, 100% isolates
Test Time 10 min.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.