Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ MeriFluor™ Chlamydia Direct Fluorescent Test Kit
GSA_VA

Catalog No. 14910112 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-910-112 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-910-112 Supplier Meridian Bioscience™ Supplier No. 500111
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Prediluted FITC-conjugated monoclonal anti-chlamydia (containing Evans blue, 0.1% sodium azide, and 1% carrier protein) and FA mounting medium

Direct fluorescent antigen detection of chlamydia in McCoy cells

  • Infected cells are visualized using a fluorescence microscope
  • Employs fluorescein (FITC) conjugated monoclonal antibodies specific for all known serotypes of Chlamydia trachomatis as well as Chlamydia psittaci
  • Positive results: 1+ or greater fluorescence; negative: no fluorescent chlamydia inclusion bodies seen
  • For in vitro diagnostic use

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Content And Storage 2°C to 8°C
Detectable Analytes Chlamydia species
Format IFA
Type Chlamydia Direct Fluorescent Test Kit
Includes Prediluted FITC-conjugated monoclonal Anti Chlamydia (containing evans blue, 0.1% Sodium Azide and 1% carrier Protein) and FA mounting medium
Quantity 50 Tests
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.