Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ MeriFluor-Pneumocystis carinii
GSA_VA

Catalog No. 23288103 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
30 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-288103 30 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-288103 Supplier Meridian Bioscience™ Supplier No. 222030
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Ready-to-use detection reagent, mounting fluid, specimen pipets and slides for 30 determinations

Kit readily detects rare organisms

  • Test results in 40 minutes
  • CLIA Complexity: High
  • Direct, immunofluorescent, detection procedure for Pneumocystis carinii cysts and trophozoites
  • Uses monoclonal antibodies to directly bind with the Pneumocystis organisms in respiratory tract specimens
  • Positive specimens are microscopically read by apple-green fluorescence

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance CE Marked
CPT Code 87281
Clia Complexity High
Sample Type Bronchial Wash, Bronchoalveolar Lavage, Induced Sputum
Type Pneumocystis carinii Test Kit
For Use With (Application) Readily detects rare organisms
Includes Ready-to-use detection reagent, mounting fluid, specimen pipets and slides for 30 determinations
Quantity 30 Tests
Test Time 40 min.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.