Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Meridian Bioscience™ MONOSPOT™ Mono Test Kit
Shop All Meridian Bioscience Products
Click to view available options
Quantity:
50 Tests
Description
Includes
Kits contain test cards, latex reagent, positive control, negative control, and Pipetstirs.
- Rapid latex test for the detection of heterophile antibodies specific to infectious mononucleosis in human serum or plasma (fresh, refrigerated, or frozen)
- Results within 5 minutes
- Positive and negative controls; ready-to-use reagents
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Detectable Analytes | Infectious Mononucleosis Heterophil Antibodies |
| Format | Kit |
| Sensitivity | 94% |
| Type | Mononucleosis Test Kit |
| Includes | Test cards, Latex reagent, positive control, negative control and pipetstirs |
| Quantity | 50 Tests |
| Specificity | 0.93 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction