Learn More
Meridian Bioscience™ Para-Pak™ EcoStain Trichrome Staining System
Shop All Meridian Bioscience ProductsDescription
Includes
Contains three components (Trichrome Stain, Trichrome Enhancer A and Trichrome B) that, when mixed together, provide good differentiation of stool parasites from background material as well as excellent staining of parasite intracellular components.
Shorter procedure than conventional trichrome; Set includes all reagents necessary.
EcoStain is a rapid staining procedure designed to stain intestinal parasites from stool that has been fixed in nonmercury-based PVA preservatives. Components generally stain purple to red; cytoplasm usually stains blue/green; stool background generally stains blue/green/purple. Hemo-De and Modified Hemo-De are provided as xylene substitutes in the dehydration and clearing steps.
Order Info
Stains 350 to 400 slides. For in vitro diagnostic use.
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| Format | Bottle |
| Sample Type | Stool |
| Type | EcoStain Trichrome Staining System |
| For Use With (Application) | Rapid staining procedure designed to stain intestinal parasites from stool and used with EcoFix™ and non-mercury PVA preservatives |
| Includes | Three components of Trichrome stain, Trichrome enhancer A and Trichrome B and necessary reagents |
| Quantity | 350 to 400 Pack |
| Test Time | 12 min. |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.