Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ Premier™ CAMPY Enzyme Immunoassay
GSA_VA

Catalog No. 23315100 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
96 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-315-100 96 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

Patient specimens may contain infectious agents and should be handled and disposed of as potential biohazards.

Dispose of used wash buffer and all test materials in an appropriate container. Treat waste as a potential biohazard.

The Positive Control reagent contains inactivated C. jejuni. It should be handled, however, as a potential biohazard.

Avoid skin contact with Premier Stop Solution I. Flush with water immediately if contact occurs.

Catalog No. 23-315-100 Supplier Meridian Bioscience™ Supplier No. 618096
 Restricted Item
Add to Cart
Add to Cart

Patient specimens may contain infectious agents and should be handled and disposed of as potential biohazards.

Dispose of used wash buffer and all test materials in an appropriate container. Treat waste as a potential biohazard.

The Positive Control reagent contains inactivated C. jejuni. It should be handled, however, as a potential biohazard.

Avoid skin contact with Premier Stop Solution I. Flush with water immediately if contact occurs.

Enzyme immunoassay for the detection of Campylobacter antigens in stool specimens

  • Optimized performance: Provides detection without lengthy and variable culture procedures
  • Faster turnaround time: Improved from 48-72 hours to just 2 hours
  • Premier™ CAMPY enzyme immunoassay (EIA) is an in vitro qualitative procedure for the detection of specific Campylobacter antigens
  • For testing of stool samples from patients with signs and symptoms of gastroenteritis
  • Premier CAMPY detects C. jejuni and C. coli in human stool
  • Test results are to be used in conjunction with information obtained clinical evaluations and other diagnostic procedures
  • Intended for use in laboratory settings
  • For in vitro diagnostic use

  • Kit includes Premier CAMPY microwells, Premier CAMPY Enzyme Conjugate, Premier 20X Wash Buffer III, Premier Substrate I, Premier CAMPY Sample Diluent/Negative Control, Premier CAMPY Positive Control, transfer pipettes, microwell strip holder and microwell plate sealers
  • Suggested CPT Code: 87449

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

CPT Code 87449
Detectable Analytes Campylobacter Antigen, C. jejuni Antigen, C. coli Antigen
Format Microwell
Sample Type Unpreserved or Cary-Blair preserved stool
Type Premier™ Campy EIA Kit
Includes Premier™ Campy microwells, Premier™ Campy Enzyme conjugate, Premier™ 20X Wash Buffer III, Premier™ substrate I, Premier™ Campy sample diluent/negative control, Premi
Quantity 96 Pack
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.