Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ Premier™ Cryptococcal Antigen
GSA_VA

Catalog No. 14910162 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
192 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-910-162 192 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-910-162 Supplier Meridian Bioscience™ Supplier No. 602096
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Capable of detecting lower antigen concentrations at early or late disease stages

  • CLIA Complexity: High
  • For the qualitative and semiquantitative detection of cryptococcal antigen in serum and cerebrospinal fluid (CSF)
  • Monoclonal EIA with test results within 30 minutes
  • More sensitive than traditional latex testing procedures
  • Sensitivity: 100%; Specificity: 98%

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Content And Storage 2°C to 8°C
CPT Code 87327
Detectable Analytes Cryptococcal Antigen (CrAg)
Sample Type CSF, Serum
Sensitivity 1
Type Premier™ Cryptococcal Antigen Test Kit
Quantity 192 Tests
Specificity 0.98
Test Time 30 min.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.