Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ Premier™ Rotaclone™ Rotavirus Test Kit
GSA_VA

Catalog No. 14910186 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
48 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-910-186 48 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-910-186 Supplier Meridian Bioscience™ Supplier No. 696004
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Kit contains enough for 48 tests: microtiter wells, conjugate, positive control, sample diluent, substrate buffer and solution, stop solution, transfer pipettes, and microtiter well holder.

EIA for the detection of Rotavirus Antigen in human fecal samples

  • Utilizes monoclonal anitbodies in a solid phase sandwich type EIA
  • Sensitivity: 100%; specificity: 100%

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance CE Marked
Content And Storage 2°C to 8°C
CPT Code 87425
Detectable Analytes Rotavirus Antigen
Format 48 Microwells
Sample Type Stool
Sensitivity 1
Type Rotavirus Test Kit
Includes Microtiter wells, conjugate, positive control, sample diluent, substrate Buffer and solution, stop solution, transfer pipettes and microtiter well holder
Quantity 48 Tests
Specificity 1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.