Learn More
Sklar Instruments Hartmann Alligator Forceps, Sklar 5-1/2 in., Blunt, Angled, Serrated, Stainless Steel. EA.
SDP
Catalog No. 501183941 Shop All Sklar Instruments Products
Encompass_Preferred
Hartmann Alligator Forceps, Sklar. 5-1/2 in., Blunt, Angled, Serrated, Stainless Steel. Merit Hartmann Alligator Forceps are an ENT instrument commonly used in myringotomy procedures to insert or remove tubes or dressings and packing. The serrated tips securely hold and remove foreign objects, granulation tissue, and cerumen. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures. These cost effective, high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee. Merit instruments deliver an economical solution for physicians and practices that require quality, durable instruments. This product is angled at the Pack lock with straight serrated tips and a length of 5-1/2 inches.
Catalog No. 50-118-3941 Supplier Sklar Instruments Supplier No. 98224
Hartmann Alligator Forceps, Sklar. 5-1/2 in., Blunt, Angled, Serrated, Stainless Steel. Merit Hartmann Alligator Forceps are an ENT instrument commonly used in myringotomy procedures to insert or remove tubes or dressings and packing. The serrated tips securely hold and remove foreign objects, granulation tissue, and cerumen. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures. These cost effective, high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee. Merit instruments deliver an economical solution for physicians and practices that require quality, durable instruments. This product is angled at the Pack lock with straight serrated tips and a length of 5-1/2 inches.