Learn More
Sklar Instruments Forceps, Merit Crile Hemostatic Forceps, Sklar 6-1/4 in., Straight, Serrated, Stainless Steel, EA.
SDP
Catalog No. 501126616 Shop All Sklar Instruments Products
Encompass_Preferred
Forceps, Merit Crile Hemostatic Forceps, Sklar, 6-1/4 in., Straight, Serrated, Stainless Steel, EA. Merit Crile Hemostatic Forceps, also known as a Crile Hemostat, are most commonly used for clamping blood vessels or tissue before cauterization or ligation. Horizontal serrations the entire length of the jaw, and jaws half the length of the shank make this instrument unique. They may also be used for soft tissue dissection typically no deeper than 6-8 inches or in Laparotomy procedures. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures. These cost effective, high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee. Merit instruments deliver an economical solution for physicians and practices that require quality, durable instruments. This product is straight with serrated tips and a length of 6-1/4 inches.
Catalog No. 50-112-6616 Supplier Sklar Instruments Supplier No. 97458
Forceps, Merit Crile Hemostatic Forceps, Sklar, 6-1/4 in., Straight, Serrated, Stainless Steel, EA. Merit Crile Hemostatic Forceps, also known as a Crile Hemostat, are most commonly used for clamping blood vessels or tissue before cauterization or ligation. Horizontal serrations the entire length of the jaw, and jaws half the length of the shank make this instrument unique. They may also be used for soft tissue dissection typically no deeper than 6-8 inches or in Laparotomy procedures. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures. These cost effective, high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee. Merit instruments deliver an economical solution for physicians and practices that require quality, durable instruments. This product is straight with serrated tips and a length of 6-1/4 inches.