Learn More
Sklar Instruments Forceps, Merit Kelly Hemostatic Forceps, Sklar 5-1/2 in., Curved, Serrated, Stainless Steel, EA.
SDP
Catalog No. 501126612 Shop All Sklar Instruments Products
Encompass_Preferred
Forceps, Merit Kelly Hemostatic Forceps, Sklar, 5-1/2 in., Curved, Serrated, Stainless Steel, EA. Merit Kelly Hemostatic Forceps are primarily used for clamping large blood vessels or manipulating heavy tissue. They may also be used for soft tissue dissection. They are available curved or straight with a ratcheted finger ring handle. The jaws of Kelly Forceps are 1/3 the length of the shanks and serrations are 1/2 the length of the jaws. Merit instruments are Sklars mid-grade line of products and most commonly utilized for routine and in-office minor procedures. This product is curved with serrated jaws and a length of 5-1/2 inches.
Catalog No. 50-112-6612 Supplier Sklar Instruments Supplier No. 97442
Forceps, Merit Kelly Hemostatic Forceps, Sklar, 5-1/2 in., Curved, Serrated, Stainless Steel, EA. Merit Kelly Hemostatic Forceps are primarily used for clamping large blood vessels or manipulating heavy tissue. They may also be used for soft tissue dissection. They are available curved or straight with a ratcheted finger ring handle. The jaws of Kelly Forceps are 1/3 the length of the shanks and serrations are 1/2 the length of the jaws. Merit instruments are Sklars mid-grade line of products and most commonly utilized for routine and in-office minor procedures. This product is curved with serrated jaws and a length of 5-1/2 inches.