Learn More
Sklar Instruments Forceps, Merit Lorna (Edna) Towel Clamps, Sklar 5-1/4 in., Non-Perforating, Straight, Serrated, Stainless Steel, EA.
SDP
Catalog No. 501126665 Shop All Sklar Instruments Products
Encompass_Preferred
Forceps, Merit Lorna (Edna) Towel Clamps, Sklar, 5-1/4 in., Non-Perforating, Straight, Serrated, Stainless Steel, EA. Merit Lorna (Edna) Towel Clamps are a non-perforating clamp used to secure instruments, surgical materials such as tubing, and towels to the drape surrounding a surgical site. The jaws and tips are curved while the shanks and handle are flat so they can rest on the patient. The teeth on the tips of this towel clamp are blunted so they grip tightly but do not penetrate the surgical drape. They also have ratcheted finger ring handles so a firm hold can be secured. The Lorna Towel Clamp is also commonly refered to as the Edna Towel Clamp. This clamp should never be used on skin. Merit instruments are Sklars mid-grade line of products and most commonly utilized for routine and in-office minor procedures. These cost effective, high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee.
Catalog No. 50-112-6665 Supplier Sklar Instruments Supplier No. 97635
Forceps, Merit Lorna (Edna) Towel Clamps, Sklar, 5-1/4 in., Non-Perforating, Straight, Serrated, Stainless Steel, EA. Merit Lorna (Edna) Towel Clamps are a non-perforating clamp used to secure instruments, surgical materials such as tubing, and towels to the drape surrounding a surgical site. The jaws and tips are curved while the shanks and handle are flat so they can rest on the patient. The teeth on the tips of this towel clamp are blunted so they grip tightly but do not penetrate the surgical drape. They also have ratcheted finger ring handles so a firm hold can be secured. The Lorna Towel Clamp is also commonly refered to as the Edna Towel Clamp. This clamp should never be used on skin. Merit instruments are Sklars mid-grade line of products and most commonly utilized for routine and in-office minor procedures. These cost effective, high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee.