Learn More
Sklar Instruments Micro Alligator Ear Forceps, Sklar 1mm Cup, Straight, Very Delicate, Stainless Steel. EA.
SDP
Catalog No. 501183942 Shop All Sklar Instruments Products
Encompass_Preferred
Micro Alligator Ear Forceps, Sklar. 1mm Cup, Straight, Very Delicate, Stainless Steel. Merit Micro Alligator Ear Forceps are non-ratcheted, finger ring forceps used for manipulating and removing tissue from the ear canal or middle ear. They may also be used for packing during grafting procedures or as nasal forceps. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures. These cost effective, high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee. Merit instruments deliver an economical solution for physicians and practices that require quality, durable instruments. This product has 2-3/4 inch straight shanks. At the Pack lock the shaft extends 5-1/2 inches to a very delicate 1mm circular cup.
Catalog No. 50-118-3942 Supplier Sklar Instruments Supplier No. 98234
Micro Alligator Ear Forceps, Sklar. 1mm Cup, Straight, Very Delicate, Stainless Steel. Merit Micro Alligator Ear Forceps are non-ratcheted, finger ring forceps used for manipulating and removing tissue from the ear canal or middle ear. They may also be used for packing during grafting procedures or as nasal forceps. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures. These cost effective, high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee. Merit instruments deliver an economical solution for physicians and practices that require quality, durable instruments. This product has 2-3/4 inch straight shanks. At the Pack lock the shaft extends 5-1/2 inches to a very delicate 1mm circular cup.