Learn More
Sklar Instruments Forceps, Merit Foerster Sponge Forceps, Sklar 7 in., Fenestrated, Straight, Serrated, Stainless Steel, EA.
SDP
Catalog No. 501126650 Shop All Sklar Instruments Products
Encompass_Preferred
Forceps, Merit Foerster Sponge Forceps, Sklar, 7 in., Fenestrated, Straight, Serrated, Stainless Steel, EA. Merit Foerster Sponge Forceps, which may also be known as Foerster-Ballenger Sponge Forceps, are commonly used in surgical procedures to hold gauze squares that will absorb excess fluids and blood from the surgical site or hemostasis. The ratcheted locking mechanism of these finger ring forceps securely grip the gauze and hold it in place. Foerster Sponge Forceps that are longer in length are often used in bariatric surgeries and the curved version may be used for removing the placenta in OB/GYN procedures. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures. These cost effective, high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee.
Catalog No. 50-112-6650 Supplier Sklar Instruments Supplier No. 97571
Forceps, Merit Foerster Sponge Forceps, Sklar, 7 in., Fenestrated, Straight, Serrated, Stainless Steel, EA. Merit Foerster Sponge Forceps, which may also be known as Foerster-Ballenger Sponge Forceps, are commonly used in surgical procedures to hold gauze squares that will absorb excess fluids and blood from the surgical site or hemostasis. The ratcheted locking mechanism of these finger ring forceps securely grip the gauze and hold it in place. Foerster Sponge Forceps that are longer in length are often used in bariatric surgeries and the curved version may be used for removing the placenta in OB/GYN procedures. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures. These cost effective, high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee.