Learn More
AdipoGen Merocyanin 540
SDP
Supplier:  AdipoGen CDXM0033M500
Encompass_Preferred
Chemical. CAS 62796-23-0. Formula C26H32N3NaO6S2. MW 569.67. Synthetic. Sensitive probe for membrane potential Ex/Em 555/578nm in methanol. Selective staining of immature hemopoietic cells in flow cytometry.
Catalog No. 50-256-0555