Learn More
AdipoGen Merocyanin 540,Chemical. CAS: 62796-23-0
SDP
Catalog No. 501177597
Encompass_Preferred
Merocyanin 540, AdipoGen Life Sciences. Chemical. CAS: 62796-23-0. Formula: C26H32N3NaO6S2. MW: 569.67. Synthetic. Sensitive probe for membrane potential (Ex/Em: 555/578nm in methanol). Selective staining of immature hemopoietic cells in flow cytometry. 62796-23-0
Catalog No. 50-117-7597 Supplier AdipoGen Supplier No. CDXM0033M100
Merocyanin 540, AdipoGen Life Sciences. Chemical. CAS: 62796-23-0. Formula: C26H32N3NaO6S2. MW: 569.67. Synthetic. Sensitive probe for membrane potential (Ex/Em: 555/578nm in methanol). Selective staining of immature hemopoietic cells in flow cytometry. 62796-23-0