Learn More
Lkt Laboratories Meropenem Sodium Carbonate, ≥98%, 25MG
SDP
Catalog No. NC9737753
Encompass_Preferred
Carbapenem β-lactam; penicillin binding protein inhibitor. CAS: 96036-03-2. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. NC9737753 Supplier Lkt Laboratories Supplier No. M177025MG
Carbapenem β-lactam; penicillin binding protein inhibitor. CAS: 96036-03-2. Additional sizes available upon request. Ships from Saint Paul, MN.