Learn More
Medchemexpress LLC Meropenem-d6 | 1217976-95-8 | 98.0% | C17H19D6N3O5S | 10MG
SDP
Catalog No. 5000259364
Encompass_Preferred
Meropenem-d6 is a deuterium-labeled isotopologue of the carbapenem antibiotic meropenem, provided as a high-purity analytical standard for research use. It is intended as an internal standard for LC-MS or GC-MS quantification and other analytical assays. Not for human or clinical use.
  • Deuterium-labeled internal standard for accurate quantification.
  • High purity (98.0%) suitable for analytical methods.
  • Compatible with LC-MS and GC-MS workflows.
  • Available in small, weighed quantities for standards preparation.
  • Intended for research use only; not for human use.
Catalog No. 50-002-59364 Supplier Medchemexpress LLC Supplier No. HY13678S10MG
Meropenem-d6 is a deuterium-labeled isotopologue of the carbapenem antibiotic meropenem, provided as a high-purity analytical standard for research use. It is intended as an internal standard for LC-MS or GC-MS quantification and other analytical assays. Not for human or clinical use.
  • Deuterium-labeled internal standard for accurate quantification.
  • High purity (98.0%) suitable for analytical methods.
  • Compatible with LC-MS and GC-MS workflows.
  • Available in small, weighed quantities for standards preparation.
  • Intended for research use only; not for human use.