Learn More
Medchemexpress LLC Meropenem-d6 (SM 7338-d6) | 1217976-95-8 | 98.3% | 389.50 | 1 MG
SDP
Catalog No. 5000350452
Encompass_Preferred
Meropenem-d6 is a deuterium-labeled version of Meropenem. Meropenem (SM 7338) is a carbapenem antibiotic known for its broad-spectrum antibacterial activity. It demonstrates activity against various susceptible and resistant bacteria, including *N. gonorrhoeae*, *H. influenzae*, and *H. ducreyi*, with reported MIC values ranging from 0.015 mg/mL to 0.12 mg/mL.
  • Used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-50452 Supplier Medchemexpress LLC Supplier No. HY13678S1MG
Meropenem-d6 is a deuterium-labeled version of Meropenem. Meropenem (SM 7338) is a carbapenem antibiotic known for its broad-spectrum antibacterial activity. It demonstrates activity against various susceptible and resistant bacteria, including *N. gonorrhoeae*, *H. influenzae*, and *H. ducreyi*, with reported MIC values ranging from 0.015 mg/mL to 0.12 mg/mL.
  • Used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.