Learn More
Biomatik Corporation Meropenem (Mer) ELISA Kit
Supplier:  Biomatik Corporation EKU12253
Encompass_Preferred
Meropenem (Mer) ELISA Kit for detecting Mer in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.94pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: Merrem. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7994