Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Meropenem trihydrate | 119478-56-7 | MFCD08600005 | 99.9% | C17H31N3O8S | 10MG
SDP

Catalog No. 5000258939
Encompass_Preferred

Meropenem trihydrate is the trihydrate form of the carbapenem antibiotic meropenem provided as an analytical reference standard for research and quality-control applications. It is intended for assay development, method validation, and instrument calibration, and is characterized by a high-purity HPLC assay suitable for analytical workflows.

  • High purity reference material (≈99.9% by RP-HPLC).
  • Suitable for HPLC method validation and system suitability.
  • Stable solid trihydrate form for consistent analytical performance.
  • Supplies small lab-scale pack sizes for reference and QC work.
  • Compatible with routine assay development and quantitative analysis.
  • Characterized by molecular formula C17H31N3O8S and CAS 119478-56-7.

Catalog No. 50-002-58939 Supplier Medchemexpress LLC Supplier No. HY13678AR10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Meropenem trihydrate is the trihydrate form of the carbapenem antibiotic meropenem provided as an analytical reference standard for research and quality-control applications. It is intended for assay development, method validation, and instrument calibration, and is characterized by a high-purity HPLC assay suitable for analytical workflows.

  • High purity reference material (≈99.9% by RP-HPLC).
  • Suitable for HPLC method validation and system suitability.
  • Stable solid trihydrate form for consistent analytical performance.
  • Supplies small lab-scale pack sizes for reference and QC work.
  • Compatible with routine assay development and quantitative analysis.
  • Characterized by molecular formula C17H31N3O8S and CAS 119478-56-7.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.