Learn More
Medchemexpress LLC Meropenem (trihydrate) (standard) | 119478-56-7 | 100.0% | 437.51 | 100 MG
SDP
Catalog No. 5000347956
Encompass_Preferred
Meropenem (trihydrate) (Standard) is the analytical standard of Meropenem (trihydrate), a carbapenem antibiotic with broad-spectrum antibacterial activity. It is effective against susceptible and resistant N. gonorrhoeae, H. influenzae, and H. ducreyi. This product is intended for research and analytical applications.
  • Analytical standard for meropenem (trihydrate).
  • Carbapenem antibiotic with broad-spectrum antibacterial activity.
  • Effective against susceptible and resistant N. gonorrhoeae, H. influenzae, and H. ducreyi.
  • Suitable for research and analytical applications, including HPLC, GC, and MS.
Catalog No. 50-003-47956 Supplier Medchemexpress LLC Supplier No. HY13678AR100MG
Meropenem (trihydrate) (Standard) is the analytical standard of Meropenem (trihydrate), a carbapenem antibiotic with broad-spectrum antibacterial activity. It is effective against susceptible and resistant N. gonorrhoeae, H. influenzae, and H. ducreyi. This product is intended for research and analytical applications.
  • Analytical standard for meropenem (trihydrate).
  • Carbapenem antibiotic with broad-spectrum antibacterial activity.
  • Effective against susceptible and resistant N. gonorrhoeae, H. influenzae, and H. ducreyi.
  • Suitable for research and analytical applications, including HPLC, GC, and MS.