Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Meropenem trihydrate (Standard) | 119478-56-7 | 100.0% | 437.51 | 50 MG
SDP

Catalog No. 5000348685
Encompass_Preferred

Meropenem trihydrate (Standard) | 119478-56-7 | 100.0% | 437.51 | 50 MG

Catalog No. 50-003-48685 Supplier Medchemexpress LLC Supplier No. HY13678AR50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Meropenem trihydrate (Standard) | 119478-56-7 | 100.0% | 437.51 | 50 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.