Learn More
Medchemexpress LLC Meropenem-vaborbactam | 2031124-72-6 | 97.9% | 680.60 | 25 MG
SDP
Catalog No. 5000346741
Encompass_Preferred
Meropenem-vaborbactam is a Carbapenem antibiotic and boronic acid-based beta-lactamase inhibitor. This fixed-dose combination product shows potent in vitro activity against Enterobacteriaceae, including Klebsiella pneumoniae carbapenemase producers. It also demonstrates activity against Pseudomonas aeruginosa isolates.
  • Potently inhibits Enterobacteriaceae isolates.
  • Exhibits activity against Pseudomonas aeruginosa isolates.
  • Demonstrates potent in vitro activity against 991 KPC-producing Enterobacterales strains.
  • Achieves bacteriostasis at an fAUC:MIC ratio of 9 in a murine model.
  • Achieves a 1-log10 bacterial kill at an fAUC:MIC ratio of 38 in a murine model.
Catalog No. 50-003-46741 Supplier Medchemexpress LLC Supplier No. 25MGHY111521
Meropenem-vaborbactam is a Carbapenem antibiotic and boronic acid-based beta-lactamase inhibitor. This fixed-dose combination product shows potent in vitro activity against Enterobacteriaceae, including Klebsiella pneumoniae carbapenemase producers. It also demonstrates activity against Pseudomonas aeruginosa isolates.
  • Potently inhibits Enterobacteriaceae isolates.
  • Exhibits activity against Pseudomonas aeruginosa isolates.
  • Demonstrates potent in vitro activity against 991 KPC-producing Enterobacterales strains.
  • Achieves bacteriostasis at an fAUC:MIC ratio of 9 in a murine model.
  • Achieves a 1-log10 bacterial kill at an fAUC:MIC ratio of 38 in a murine model.