Learn More
Cayman Chemical Meropenmhydrate 100mg
SDP
Catalog No. 502972331
Encompass_Preferred
A carbapenem antibiotic; active against clinical isolates of Gram-positive and Gram-negative bacteria in vitro, including S. pneumoniae, H. influenzae, N. gonorrhoeae, E. coli, P. aeruginosa, C. difficile, and methicillin-susceptible and -resistant S. aureus (MICs = ≤0.008-8 μg/ml); protective against S. aureus, S. pneumoniae, E. coli, S. marcescens, P. mirabilis, and P. aeruginosa infections in mice (PD50s = 0.13, 0.01, 0.04, 0.07, 0.84, and 0.46 mg/kg, respectively)
Catalog No. 50-297-2331 Supplier Cayman Chemical Supplier No. 16068100 MG
A carbapenem antibiotic; active against clinical isolates of Gram-positive and Gram-negative bacteria in vitro, including S. pneumoniae, H. influenzae, N. gonorrhoeae, E. coli, P. aeruginosa, C. difficile, and methicillin-susceptible and -resistant S. aureus (MICs = ≤0.008-8 μg/ml); protective against S. aureus, S. pneumoniae, E. coli, S. marcescens, P. mirabilis, and P. aeruginosa infections in mice (PD50s = 0.13, 0.01, 0.04, 0.07, 0.84, and 0.46 mg/kg, respectively)