Learn More
Sino Biological MERS-CoV NP-CoV PAb 20ul (Sino Biological)
Supplier:  Sino Biological 100213RP0220
Encompass_Preferred
Produced in rabbits immunized with purified, a synthetic peptide corresponding to the center region of MERS-CoV (NCoV / Novel coronavirus) Nucleocapsid Protein (NP protein), and purified by antigen affinity chromatography.
Catalog No. 50-237-2297