Learn More
Enzo Life Sciences Coronavirus (MERS-CoV S1) antigen (aa 56-295), (recombinant) (His-tag) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2970100
Encompass_Preferred
Alternative name: Coronavirus spike protein. Purity: ≥95%. Formulation: Liquid. In 10 mM PBS, pH 7.2 with 25 mM arginine and 0.05% sodium azide. Source: Produced in E. coli. Coronavirus (MERS-CoV) spike protein (aa 56-295) is fused to a His-tag. Long Term Storage: -20°C. Applications: ELISA .
Catalog No. 50-201-4108