Learn More
Sino Biological MERS-CoV Spike Protein 20ug (Sino Biological)
Supplier:  Sino Biological 40069V27HB20
Encompass_Preferred
A DNA sequence encoding the MERS-CoV spike (AFS88936.1) (Glu367-Tyr606) was expressed with a c-terminal AVI tagged polyhistidine tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-1321