Learn More
ACROBiosystems Biotinylated MERS Spike Trimer Protein, His,Avitag™ (MALS verified)
Catalog No. 502331403
Encompass_Preferred
Biotinylated MERS Spike Trimer Protein, His,Avitag (SPN-M82E3) is expressed from human 293 cells (HEK293). It contains AA Tyr 18 - Trp 1295 (Accession # K9N5Q8-1 (R748A, R751A, V1060P, L1061P, K1284Q)).
Catalog No. 50-233-1403 Supplier ACROBiosystems Supplier No. SPNM82E325UG
Biotinylated MERS Spike Trimer Protein, His,Avitag (SPN-M82E3) is expressed from human 293 cells (HEK293). It contains AA Tyr 18 - Trp 1295 (Accession # K9N5Q8-1 (R748A, R751A, V1060P, L1061P, K1284Q)).