Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mersalyl acid, ≥85% (TLC), MilliporeSigma™ Supelco™

Catalog No. 111017777 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7777 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7777 Supplier MilliporeSigma Supelco Supplier No. M97841G
Add to Cart
Add to Cart
Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Packaging Glass Bottle
CAS 486-67-9
Physical Form Neat
Linear Formula HOHgCH2 CH(OCH3)CH2 NHCOC6H4 OCH2 CO2 H
MDL Number MFCD00004302
Melting Point 192°C to 193°C
Synonym 2-(3-Hydroxymercurio-2-methoxypropylcarbamoyl)phenoxyacetic acid; 2-[N-(3-Hydroxymercuri-2-methoxypropyl)carbamoyl]phenoxyacetic acid; Salyrganic acid
Solubility Soluble in NH4OH
Type Analytical Standard
Quantity 1 g
Formula Weight 483.87
Percent Purity ≥85% (TLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.