Learn More
Sino Biological MERS-CoV Nucleocapsid Antibody, Mouse MAb
Supplier:  Sino Biological 40068MM10100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant MERS-CoV (NCoV / Novel coronavirus) Nucleoprotein / NP (Catalog#40068-V08B; AFS88943.1; Met 1-Asp413). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-228-2315