Learn More
Sino Biological MERS-CoV Spike S1+S2 Protein (ECD, aa 1-1297, His Tag), Biotinylated
Supplier:  Sino Biological 40069V08BB20
Encompass_Preferred
A DNA sequence encoding the MERS-CoV extracellular domain of spike protein (AFS88936.1) (Met1-Trp1297) was fused with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-224-7179