Learn More
Medchemexpress LLC HY-19792 50mg , Mertansine CAS:139504-50-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1979250MG
Encompass_Preferred
HY-19792 50mg Mertansine CAS: 139504-50-0 Mertansine (DM1) is a microtubulin inhibitor and is an antibody-conjugatable maytansinoid that is developed to overcome systemic toxicity associated with maytansine and to enhance tumor-specific delivery. Mertansine can be attached to a monoclonal antibody with a linker to create an antibody-drug conjugate (ADC). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9155