Learn More
ACROBiosystems ACROBiosystems Human MERTK / Mer Protein, His Tag
Catalog No. 501956730
Encompass_Preferred
Recombinant Protein;1MG;Human MERTK, His Tag (MEK-H52H6) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Ile 505 (Accession # Q12866-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 54.5 kDa. The protein migrates as 70-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6730 Supplier ACROBiosystems Supplier No. MEKH52H61MG
Recombinant Protein;1MG;Human MERTK, His Tag (MEK-H52H6) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Ile 505 (Accession # Q12866-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 54.5 kDa. The protein migrates as 70-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.