Learn More
MSC Koch Filter Pleated Air Filter, MERV 8, High Capacity, 6 in x 24 in x 24 in, 1/ea
Catalog No. 5000507215
Encompass
The Multi-Pleat XL8-HC is a high-capacity, extended-surface pleated panel filter designed for applications requiring superior indoor air quality, such as hospitals, laboratories, and commercial buildings. Constructed with 100% synthetic mechanical media, this filter provides high dust holding capacity and low resistance to airflow. It is an ideal upgrade from standard disposable, MERV 6, or MERV 7 pleated filters.
  • Maintains MERV 8 and MERV 8-A efficiency per ASHRAE 52.2-2017 standards
  • Features high-strength, moisture-resistant beverage board frame with double-wall design
  • Includes expanded metal grid on downstream side for pleat uniformity
  • Suitable for continuous operation up to 200°F
  • Certified to UL 900 and ULC-S111-07 standards
  • Manufactured by an ISO 9001:2015 registered company
Catalog No. 50-005-07215 Supplier MSC Supplier No. 86093978
The Multi-Pleat XL8-HC is a high-capacity, extended-surface pleated panel filter designed for applications requiring superior indoor air quality, such as hospitals, laboratories, and commercial buildings. Constructed with 100% synthetic mechanical media, this filter provides high dust holding capacity and low resistance to airflow. It is an ideal upgrade from standard disposable, MERV 6, or MERV 7 pleated filters.
  • Maintains MERV 8 and MERV 8-A efficiency per ASHRAE 52.2-2017 standards
  • Features high-strength, moisture-resistant beverage board frame with double-wall design
  • Includes expanded metal grid on downstream side for pleat uniformity
  • Suitable for continuous operation up to 200°F
  • Certified to UL 900 and ULC-S111-07 standards
  • Manufactured by an ISO 9001:2015 registered company